Learn More
Biomatik Corporation Total MMP-8 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51990
Encompass_Preferred
Total MMP-8 (Human) Sandwich ELISA Kit for detecting Total MMP-8 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22894. Gene ID: 4317. Target Synonyms: Neutrophil collagenase; Matrix metalloproteinase-8; PMNL collagenase; CLG1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1919