Learn More
Biomatik Corporation Angiopoietin-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52011
Encompass_Preferred
Angiopoietin-2 (Human) Sandwich ELISA Kit for detecting Angiopoietin-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 94-6000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15123. Gene ID: 285. Target Synonyms: ANG-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1938