Learn More
Biomatik Corporation CD163 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52023
Encompass_Preferred
CD163 (Human) Sandwich ELISA Kit for detecting CD163 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q86VB7. Gene ID: 9332. Target Synonyms: Scavenger receptor cysteine-rich type 1 protein M130; Hemoglobin scavenger receptor; M130. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1948