Learn More
Biomatik Corporation Clusterin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52026
Encompass_Preferred
Clusterin (Human) Sandwich ELISA Kit for detecting Clusterin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10909. Gene ID: 1191. Target Synonyms: Aging-associated gene 4 protein; Apolipoprotein J; Complement cytolysis inhibitor; Complement-associated protein SP-40,40; Ku70-binding protein 1; NA1/NA2; Testosterone-repressed prostate message 2; APOJ; CLI; KUB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1950