Learn More
Biomatik Corporation ENA-78 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52034
Encompass_Preferred
ENA-78 (Human) Sandwich ELISA Kit for detecting ENA-78 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42830. Gene ID: 6374. Target Synonyms: Epithelial-derived neutrophil-activating protein 78; Neutrophil-activating peptide ENA-78; Small-inducible cytokine B5; CXCL5; SCYB5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1955