Learn More
Biomatik Corporation Endostatin (Human) Sandwich ELISA Kit
Catalog No. 502291960
Encompass_Preferred
Endostatin (Human) Sandwich ELISA Kit for detecting Endostatin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P39060; P39059. Gene ID: 80781; 1306. Target Synonyms: Collagen alpha-1(XVIII) chain; Collagen alpha-1(XV) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1960 Supplier Biomatik Corporation Supplier No. EKA52039
Endostatin (Human) Sandwich ELISA Kit for detecting Endostatin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P39060; P39059. Gene ID: 80781; 1306. Target Synonyms: Collagen alpha-1(XVIII) chain; Collagen alpha-1(XV) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.