Learn More
Biomatik Corporation HGF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52045
Encompass_Preferred
HGF (Human) Sandwich ELISA Kit for detecting HGF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08581. Gene ID: 4233. Target Synonyms: Hepatocyte growth factor receptor; HGF receptor; Proto-oncogene c-Met; Scatter factor receptor; Tyrosine-protein kinase Met. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1963