Learn More
Biomatik Corporation HVEM-L (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52049
Encompass_Preferred
HVEM-L (Human) Sandwich ELISA Kit for detecting HVEM-L in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43557. Gene ID: 8740. Target Synonyms: Tumor necrosis factor ligand superfamily member 14; Herpes virus entry mediator ligand; HVEM-L; CD258. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1967