Learn More
Biomatik Corporation MIF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52053
Encompass_Preferred
MIF (Human) Sandwich ELISA Kit for detecting MIF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14174. Gene ID: 4282. Target Synonyms: Macrophage migration inhibitory factor; Glycosylation-inhibiting factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1970