Learn More
Biomatik Corporation uPAR (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52076
Encompass_Preferred
uPAR (Human) Sandwich ELISA Kit for detecting uPAR in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q03405. Gene ID: 5329. Target Synonyms: Urokinase plasminogen activator surface receptor; U-PAR; Monocyte activation antigen Mo3; CD87; MO3; UPAR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1981