Learn More
Biomatik Corporation Betacellulin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51822
Encompass_Preferred
Betacellulin (Human) Sandwich ELISA Kit for detecting Betacellulin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35070. Gene ID: 685. Target Synonyms: BTC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1809