Learn More
Biomatik Corporation BMP-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51824
Encompass_Preferred
BMP-2 (Human) Sandwich ELISA Kit for detecting BMP-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P12643. Gene ID: 650. Target Synonyms: Bone morphogenetic protein 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1811