Learn More
Biomatik Corporation IL-3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51852
Encompass_Preferred
IL-3 (Human) Sandwich ELISA Kit for detecting IL-3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08700. Gene ID: 3562. Target Synonyms: Interleukin-3; Hematopoietic growth factor; Mast cell growth factor; Multipotential colony-stimulating factor; P-cell-stimulating factor; MCGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1828