Learn More
Biomatik Corporation PLGF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51873
Encompass_Preferred
PLGF (Human) Sandwich ELISA Kit for detecting PLGF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49763. Gene ID: 5228. Target Synonyms: Placenta growth factor; PGFL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1841