Learn More
Biomatik Corporation IFN-gamma (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51892
Encompass_Preferred
IFN-gamma (Murine) Sandwich ELISA Kit for detecting IFN-gamma in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 23-1500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01580. Gene ID: 15978. Target Synonyms: Interferon gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1856