Learn More
Biomatik Corporation IL-2 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51900
Encompass_Preferred
IL-2 (Murine) Sandwich ELISA Kit for detecting IL-2 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 16-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04351. Gene ID: 16183. Target Synonyms: Interleukin-2; T-cell growth factor; TCGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1862