Learn More
Biomatik Corporation MIP-1alpha (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51911
Encompass_Preferred
MIP-1alpha (Murine) Sandwich ELISA Kit for detecting MIP-1alpha in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10855. Gene ID: 20302. Target Synonyms: C-C motif chemokine 3,Heparin-binding chemotaxis protein; L2G25B; Macrophage inflammatory protein 1-alpha; SIS-alpha; Small-inducible cytokine A3; TY-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1868