Learn More
Biomatik Corporation MIP-2 (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51913
Encompass_Preferred
MIP-2 (Murine) Sandwich ELISA Kit for detecting MIP-2 in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10889. Gene ID: 20310. Target Synonyms: C-X-C motif chemokine 2; Macrophage inflammatory protein 2; Scyb2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1870