Learn More
Biomatik Corporation IL-1alpha (Rat) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51922
Encompass_Preferred
IL-1alpha (Rat) Sandwich ELISA Kit for detecting IL-1alpha in plasma, serum, lysate. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16598. Gene ID: 24493. Target Synonyms: Interleukin-1 alpha; IL-1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1877