Learn More
Biomatik Corporation G-CSFR (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51940
Encompass_Preferred
G-CSFR (Human) Sandwich ELISA Kit for detecting G-CSFR in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99062. Gene ID: 1441. Target Synonyms: Granulocyte colony-stimulating factor receptor; G-CSF receptor; CD114; GCSFR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1887