Learn More
Biomatik Corporation IL-13 Ralpha1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51947
Encompass_Preferred
IL-13 Ralpha1 (Human) Sandwich ELISA Kit for detecting IL-13 Ralpha1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 250-16000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P78552. Gene ID: 3597. Target Synonyms: Interleukin-13 receptor subunit alpha-1; Cancer/testis antigen 19; CD213a1; IL-13RA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1892