Learn More
Biomatik Corporation IL-17R (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51951
Encompass_Preferred
IL-17R (Human) Sandwich ELISA Kit for detecting IL-17R in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96F46. Gene ID: 23765. Target Synonyms: Interleukin-17 receptor A; CDw217. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1895