Learn More
Biomatik Corporation IL-6R (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51965
Encompass_Preferred
IL-6R (Human) Sandwich ELISA Kit for detecting IL-6R in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08887. Gene ID: 3570. Target Synonyms: Interleukin-6 receptor subunit alpha; IL-6R 1; Membrane glycoprotein 80; CD126. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1903