Learn More
Biomatik Corporation P-Cadherin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51974
Encompass_Preferred
P-Cadherin (Human) Sandwich ELISA Kit for detecting P-Cadherin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22223. Gene ID: 1001. Target Synonyms: Placental cadherin; Cadherin-3; CDHP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1909