Learn More
Biomatik Corporation VEGF-R3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51977
Encompass_Preferred
VEGF-R3 (Human) Sandwich ELISA Kit for detecting VEGF-R3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35916. Gene ID: 2324. Target Synonyms: Vascular endothelial growth factor receptor 3; Fms-like tyrosine kinase 4; Tyrosine-protein kinase receptor FLT4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1912