Learn More
Biomatik Corporation TIMP-4 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51985
Encompass_Preferred
TIMP-4 (Human) Sandwich ELISA Kit for detecting TIMP-4 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99727. Gene ID: 7079. Target Synonyms: Metalloproteinase inhibitor 4; Tissue inhibitor of metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1915