Learn More
Biomatik Corporation Total MMP-3 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51988
Encompass_Preferred
Total MMP-3 (Human) Sandwich ELISA Kit for detecting Total MMP-3 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08254. Gene ID: 4314. Target Synonyms: Stromelysin-1; Matrix metalloproteinase-3; STMY1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1917