Learn More
Biomatik Corporation IL-17F (Murine) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51998
Encompass_Preferred
IL-17F (Murine) Sandwich ELISA Kit for detecting IL-17F in plasma, serum, lysate. Species Reactivity: Mouse (Mus musculus). Detection Range: 24-1500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q7TNI7. Gene ID: 257630. Target Synonyms: Interleukin-17F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1926