Learn More
Biomatik Corporation Angiogenin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52009
Encompass_Preferred
Angiogenin (Human) Sandwich ELISA Kit for detecting Angiogenin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P03950. Gene ID: 283. Target Synonyms: Ribonuclease 5; RNASE5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1936