Learn More
Biomatik Corporation CD23 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52024
Encompass_Preferred
CD23 (Human) Sandwich ELISA Kit for detecting CD23 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06734. Gene ID: 2208. Target Synonyms: Low affinity immunoglobulin epsilon Fc receptor; BLAST-2; C-type lectin domain family 4 member J; Fc-epsilon-RII; Immunoglobulin E-binding factor; Lymphocyte IgE receptor; CD23A; CLEC4J; FCE2; IGEBFCD23A; CLEC4J; FCE2; IGEBF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1949