Learn More
Biomatik Corporation Complement Factor D (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52028
Encompass_Preferred
Complement Factor D (Human) Sandwich ELISA Kit for detecting Complement Factor D in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P00746. Gene ID: 1675. Target Synonyms: Adipsin; C3 convertase activator; Properdin factor D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1952