Learn More
Biomatik Corporation DPPIV (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52038
Encompass_Preferred
DPPIV (Human) Sandwich ELISA Kit for detecting DPPIV in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27487. Gene ID: 1803. Target Synonyms: Dipeptidyl peptidase 4; ADABP; Adenosine deaminase complexing protein 2; ADCP-2; Dipeptidyl peptidase IV; DPP IV; T-cell activation antigen CD26; TP103. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1959