Learn More
Biomatik Corporation MBL (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52051
Encompass_Preferred
MBL (Human) Sandwich ELISA Kit for detecting MBL in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11226. Gene ID: 4153. Target Synonyms: Mannose-binding protein C; Collectin-1; MBP1; Mannan-binding protein; Mannose-binding lectin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1969