Learn More
Biomatik Corporation PDGF-AB (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52056
Encompass_Preferred
PDGF-AB (Human) Sandwich ELISA Kit for detecting PDGF-AB in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 16-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04085; P01127. Gene ID: 5154; 5155. Target Synonyms: Platelet-derived growth factor subunit A/B; PDGF subunit A/B; PDGF-1/2; Platelet-derived growth factor A/B chain; Platelet-derived growth factor alpha polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1972