Learn More
Biomatik Corporation Progranulin (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52059
Encompass_Preferred
Progranulin (Human) Sandwich ELISA Kit for detecting Progranulin in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 63-4000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P28799. Gene ID: 2896. Target Synonyms: Granulins; Proepithelin; PEPI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1974