Learn More
Biomatik Corporation Tie-1 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52072
Encompass_Preferred
Tie-1 (Human) Sandwich ELISA Kit for detecting Tie-1 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 157-10000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35590. Gene ID: 7075. Target Synonyms: Tyrosine-protein kinase receptor Tie-1; TIE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1978