Learn More
Biomatik Corporation Human Abl Interactor 1 (ABI1) ELISA Kit
Supplier:  Biomatik Corporation EKU02064
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8IZP0. Target Synonyms: ABI-1; E3B1; NAP1BP; SSH3BP1; Spectrin SH3 Domain Binding Protein 1; Abelson interactor 1; Abl-binding protein 4; Nap1-binding protein; Eps8 SH3 domain-binding protein. Restriction: For Research Use Only.
Catalog No. 50-229-2050