Learn More
Biomatik Corporation Rat Activity Regulated Cytoskeleton Associated Protein (ARC) ELISA Kit
Supplier:  Biomatik Corporation EKU02120
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q63053. Target Synonyms: Arg3.1; Activity-regulated gene 3.1 protein homolog. Restriction: For Research Use Only.
Catalog No. 50-229-2057