Learn More
Biomatik Corporation Human AF4/FMR2 Family, Member 1 (AFF1) ELISA Kit
Supplier:  Biomatik Corporation EKU02197
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.064ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P51825. Target Synonyms: AF-4; AF4; AF4-MLL; MLL/AF4; MLLT2; PBM1; Myeloid/Lymphoid Or Mixed-Lineage Leukemia; Translocated To 2; Pre-B-Cell Monocytic Leukemia Partner 1. Restriction: For Research Use Only.
Catalog No. 50-229-2072