Learn More
Biomatik Corporation Simian Alpha-Fetoprotein (AFP) ELISA Kit
Supplier:  Biomatik Corporation EKU02295
Encompass_Preferred
Simian Alpha-Fetoprotein (AFP) ELISA Kit for detecting AFP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F6VWH6. Gene ID: 704786. Target Synonyms: aFP; A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2087