Learn More
Biomatik Corporation Rat Angiopoietin Like Protein 2 (ANGPTL2) ELISA Kit
Supplier:  Biomatik Corporation EKU02370
Encompass_Preferred
Rat Angiopoietin Like Protein 2 (ANGPTL2) ELISA Kit for detecting ANGPTL2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JJ03. Gene ID: 171100. Target Synonyms: HARP; ARP2; Angiopoietin-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2103