Learn More
Biomatik Corporation Rat Angiotensin I (AngI) ELISA Kit
Supplier:  Biomatik Corporation EKU02392
Encompass_Preferred
Rat Angiotensin I (AngI) ELISA Kit for detecting AngI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1000pg/ml. Sensitivity: 5.15pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01015. Gene ID: 24179. Target Synonyms: Ang-I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2106