Learn More
Biomatik Corporation Human Antigen Peptide Transporter 1 (TAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU02449
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.134ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q03518. Target Synonyms: APT1; ABC17; ABCB2; PSF1; RING4; Y3; TAP1N; Peptide Supply Factor 1; Atp-Binding Cassette,Subfamily B,Member 2; Peptide transporter involved in antigen processing 1. Restriction: For Research Use Only.
Catalog No. 50-229-2113