Learn More
Biomatik Corporation Bovine Apolipoprotein B100 (APOB100) ELISA Kit
Supplier:  Biomatik Corporation EKU02502
Encompass_Preferred
Bovine Apolipoprotein B100 (APOB100) ELISA Kit for detecting APOB100 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 18.8-1,200ng/ml. Sensitivity: 7.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E1BNR0. Target Synonyms: Apo-B100. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2124