Learn More
Biomatik Corporation Mouse Arrestin Beta 1 (ARRb1) ELISA Kit
Supplier:  Biomatik Corporation EKU02573
Encompass_Preferred
Mouse Arrestin Beta 1 (ARRb1) ELISA Kit for detecting ARRb1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BWG8. Gene ID: 109689. Target Synonyms: ARB1; ARR1; ARR-B1; Arrestin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2132