Learn More
Biomatik Corporation Human Asc Type Amino Acid Transporter 1 (ASC1) ELISA Kit
Supplier:  Biomatik Corporation EKU02586
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC7A10; Solute Carrier Family 7 Member 10,Neutral Amino Acid Transporter,y+ System. Restriction: For Research Use Only.
Catalog No. 50-229-2135