Learn More
Biomatik Corporation Human B-Cell Activation Factor Receptor (BAFFR) ELISA Kit
Supplier:  Biomatik Corporation EKU02667
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.060ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q96RJ3. Target Synonyms: CD268; TNFRSF13C; TNFRSF13-C; BAFF-R; BAFFR; BR3; Tumor Necrosis Factor Receptor Superfamily Member 13C. Restriction: For Research Use Only.
Catalog No. 50-229-2146