Learn More
Biomatik Corporation Human Bcl2 Antagonist/Killer 1 (BAK1) ELISA Kit
Supplier:  Biomatik Corporation EKU02676
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.625-40ng/mL. Sensitivity: 0.274ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: BAK; BAK-LIKE; BCL2L7; CDN1; Bcl-2 Homologous Antagonist/Killer; Apoptosis regulator BAK; Bcl-2-like protein 7. Restriction: For Research Use Only.
Catalog No. 50-229-2148