Learn More
Biomatik Corporation Rat Beclin 1 (BECN1) ELISA Kit
Supplier:  Biomatik Corporation EKU02685
Encompass_Preferred
Rat Beclin 1 (BECN1) ELISA Kit for detecting BECN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91XJ1. Gene ID: 114558. Target Synonyms: ATG6; VPS30; Coiled-Coil,Myosin-Like BCL2 Interacting Protein; ATG6 Autophagy Related 6 Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2150