Learn More
Biomatik Corporation Human Beta-Site APP Cleaving Enzyme 1 (bACE1) ELISA Kit
Supplier:  Biomatik Corporation EKU02709
Encompass_Preferred
Human Beta-Site APP Cleaving Enzyme 1 (bACE1) ELISA Kit for detecting bACE1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56817. Gene ID: 23621. Target Synonyms: ASP2; Beta Secretase; Memapsin 2; Aspartyl protease 2; Beta-site amyloid precursor protein cleaving enzyme 1; Membrane-associated aspartic protease 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2154